Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srh-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073683.3 | NP_506084.2 | 6 - 7 | 1051 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |