Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276941.4 | NP_001263870.1 | 5 - 6 | 778 | 79 | |
NM_001380784.1 | NP_001367281.1 | 5 - 6 | 743 | 79 | ||
NM_001380785.1 | NP_001366791.1 | 5 - 6 | 756 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |