Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdr-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073712.6 | NP_506113.1 | 2 - 3 | 247 | 103 | |
GenBank mRNA | AY728060.1 | - | 2 - 3 | 267 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |