Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269489.4 | NP_001256418.1 | 3 - 4 | 574 | 75 | |
NM_001269490.3 | NP_001256419.1 | 3 - 4 | 382 | 75 | ||
NM_001269491.2 | NP_001256420.1 | 3 - 4 | 574 | 75 | ||
GenBank mRNA | U32326.1 | - | 3 - 4 | 580 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |