Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269494.3 | NP_001256423.1 | 3 - 4 | 646 | 72 | |
NM_073726.6 | NP_506127.1 | 4 - 5 | 862 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |