Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srt-39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047710.1 | NP_001041175.1 | 1 - 2 | 41 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |