Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpa-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073759.3 | NP_506160.1 | 13 - 14 | 1460 | 66 | |
GenBank mRNA | AY008131.1 | - | 9 - 10 | 1029 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |