Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srbc-43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047739.1 | NP_001041204.1 | 4 - 5 | 742 | 117 | |
NR_003179.1 | 2 - 3 | 290 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |