Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srbc-45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073803.2 | NP_506204.2 | 1 - 2 | 102 | 165 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |