Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bus-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047697.5 | NP_001041162.1 | 7 - 8 | 913 | 69 | |
NR_003174.1 | 7 - 8 | 959 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |