Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073845.5 | NP_506246.1 | 2 - 3 | 238 | 107 | |
GenBank mRNA | AF042069.1 | - | 2 - 3 | 238 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |