Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: shw-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001368471.3 | NP_001355445.1 | 3 - 4 | 261 | 135 | |
NM_001380790.1 | NP_001368707.1 | 3 - 4 | 252 | 135 | ||
NM_001380791.1 | NP_001366842.1 | 3 - 4 | 286 | 135 | ||
GenBank mRNA | DQ185514.1 | - | 2 - 3 | 198 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |