Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: try-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269546.3 | NP_001256475.1 | 8 - 9 | 886 | 98 | |
NM_001269547.3 | NP_001256476.1 | 2 - 3 | 138 | 98 | ||
NM_001368472.3 | NP_001355438.1 | 7 | 684 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |