Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: uig-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269559.3 | NP_001256488.1 | 3 - 4 | 422 | 68 | |
NM_001269560.3 | NP_001256489.1 | 3 - 4 | 464 | 68 | ||
NM_001380797.1 | NP_001367249.1 | 3 - 4 | 404 | 68 | ||
NM_001380798.1 | NP_001366647.1 | 3 - 4 | 556 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |