Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tag-65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073985.6 | NP_506386.1 | 2 - 3 | 244 | 153 | |
GenBank mRNA | AF536544.1 | - | 2 - 3 | 244 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |