Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tag-65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_073985.6 | NP_506386.1 | 9 - 10 | 1885 | 89 | |
GenBank mRNA | AF536544.1 | - | 9 - 10 | 1885 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |