Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: swan-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074016.8 | NP_506417.1 | 5 - 6 | 1302 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |