Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sptl-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269618.3 | NP_001256547.1 | 9 - 10 | 1366 | 148 | |
NM_001269619.2 | NP_001256548.1 | 8 - 9 | 1349 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |