Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vps-39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047698.4 | NP_001041163.1 | 1 - 2 | 95 | 70 | |
NM_001047699.5 | NP_001041164.1 | 1 - 2 | 95 | 70 | ||
NM_001129538.5 | NP_001123010.1 | 1 - 2 | 101 | 70 | ||
NM_001373154.2 | NP_001360569.1 | 1 - 2 | 101 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |