Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vps-39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047698.4 | NP_001041163.1 | 8 - 9 | 2001 | 94 | |
NM_001129538.5 | NP_001123010.1 | 8 - 9 | 1998 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |