Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trm-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380809.2 | NP_001366835.1 | 6 - 7 | 1478 | 70 | |
GenBank mRNA | AF059608.1 | - | 5 - 6 | 1338 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |