Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: set-31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074168.7 | NP_506569.1 | 8 - 9 | 1445 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |