Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bmk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269657.2 | NP_001256586.1 | 4 - 5 | 2139 | 67 | |
NM_001269658.3 | NP_001256587.1 | 6 - 7 | 2198 | 67 | ||
GenBank mRNA | AB024428.2 | - | 4 - 5 | 2145 | 67 | |
AF295093.1 | - | 4 - 5 | 2130 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |