Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gar-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029064.4 | NP_001024235.1 | 10 - 11 | 1811 | 150 | |
NM_001029065.5 | NP_001024236.1 | 9 - 10 | 1744 | 150 | ||
GenBank mRNA | AF139093.2 | - | 10 - 11 | 1833 | 150 | |
AY221631.1 | - | 9 - 10 | 1744 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |