Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gar-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029064.4 | NP_001024235.1 | 6 - 7 | 950 | 141 | |
GenBank mRNA | AF139093.2 | - | 6 - 7 | 972 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Caenorhabditis elegans Probe spans exons |