Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gar-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029064.4 | NP_001024235.1 | 7 - 8 | 1265 | 83 | |
NM_001029065.5 | NP_001024236.1 | 6 - 7 | 1198 | 83 | ||
GenBank mRNA | AF139093.2 | - | 7 - 8 | 1287 | 83 | |
AY221631.1 | - | 6 - 7 | 1198 | 83 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |