Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pmp-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269678.3 | NP_001256607.1 | 6 - 7 | 1424 | 104 | |
NM_001269679.3 | NP_001256608.1 | 5 - 6 | 1326 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |