Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pmp-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269678.3 | NP_001256607.1 | 7 - 8 | 1985 | 80 | |
NM_001269679.3 | NP_001256608.1 | 6 - 7 | 1887 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Inventoried Probe spans exons |