Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: set-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269683.3 | NP_001256612.1 | 14 - 15 | 4047 | 158 | |
NM_001269684.3 | NP_001256613.1 | 13 - 14 | 3368 | 158 | ||
NM_001269685.3 | NP_001256614.1 | 13 - 14 | 3359 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |