Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hil-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074279.6 | NP_506680.1 | 4 - 5 | 455 | 70 | |
GenBank mRNA | AF002679.1 | - | 4 - 5 | 428 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |