Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: xbx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074282.4 | NP_506683.2 | 1 - 2 | 225 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |