Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cng-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129446.4 | NP_001122918.1 | 6 - 7 | 1450 | 99 | |
NR_070019.1 | 7 - 8 | 1245 | 99 | |||
NR_070020.1 | 6 - 7 | 1122 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |