Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srw-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269730.1 | NP_001256659.1 | 1 - 2 | 591 | 87 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |