Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srz-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074393.4 | NP_506794.3 | 1 - 2 | 600 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |