Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: str-27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322595.1 | NP_001309461.1 | 4 - 5 | 772 | 99 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |