Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cgt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074570.5 | NP_506971.2 | 7 - 8 | 1355 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |