Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074585.9 | NP_506986.1 | 2 - 3 | 293 | 87 | |
GenBank mRNA | AF301606.1 | - | 2 - 3 | 241 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |