Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074614.8 | NP_507015.3 | 6 - 7 | 1003 | 59 | |
NR_136352.1 | 4 - 5 | 884 | 59 | |||
NR_136353.1 | 6 - 7 | 935 | 59 | |||
NR_136354.1 | 6 - 7 | 959 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |