Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srx-43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074621.3 | NP_507022.2 | 5 - 6 | 711 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |