Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: str-97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074667.3 | NP_507068.3 | 2 - 3 | 279 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |