Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074689.6 | NP_507090.1 | 1 - 2 | 242 | 116 | |
GenBank mRNA | U14524.1 | - | 1 - 2 | 123 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |