Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srbc-83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074715.2 | NP_507116.2 | 1 - 2 | 638 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |