Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pptr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074732.6 | NP_507133.4 | 7 - 8 | 1355 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |