Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dkf-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129550.2 | NP_001123022.1 | 7 - 8 | 1169 | 82 | |
NM_001269795.3 | NP_001256724.1 | 12 - 13 | 1704 | 82 | ||
NM_001269796.3 | NP_001256725.1 | 13 - 14 | 1785 | 82 | ||
NM_001269797.3 | NP_001256726.1 | 8 - 9 | 1044 | 82 | ||
NM_074838.6 | NP_507239.2 | 12 - 13 | 1773 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |