Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dkf-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129550.2 | NP_001123022.1 | 12 - 13 | 2538 | 76 | |
NM_001269795.3 | NP_001256724.1 | 17 - 18 | 3073 | 76 | ||
NM_001269796.3 | NP_001256725.1 | 18 - 19 | 3154 | 76 | ||
NM_001269797.3 | NP_001256726.1 | 13 - 14 | 2413 | 76 | ||
NM_001356774.3 | NP_001343593.1 | 4 - 5 | 1483 | 76 | ||
NM_074838.6 | NP_507239.2 | 17 - 18 | 3142 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Caenorhabditis elegans Probe spans exons |