Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec-35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269798.2 | NP_001256727.1 | 5 - 6 | 565 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |