Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec-31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074857.3 | NP_507258.2 | 5 - 6 | 726 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |