Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: par-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380857.1 | NP_001366977.1 | 9 - 10 | 1374 | 73 | |
NM_001380858.1 | NP_001367285.1 | 9 - 10 | 1351 | 73 | ||
NM_001380859.1 | NP_001367284.1 | 6 - 7 | 940 | 73 | ||
GenBank mRNA | AF160189.1 | - | 9 - 10 | 1397 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |