Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srj-26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075203.1 | NP_507604.1 | 4 - 5 | 556 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |