Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttr-24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269883.3 | NP_001256812.1 | 2 - 3 | 379 | 122 | |
NM_001269884.3 | NP_001256813.1 | 1 - 2 | 115 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |